Product Details

SNP ID
rs3775726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:157077644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGTTAAACCATGCTTCCAGATA[C/T]CTTTCTGAACATATTTTTGATATAC
Phenotype
MIM: 138492
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLRB PubMed Links

Gene Details

Gene
GLRB
Gene Name
glycine receptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000824.4 Intron NP_000815.1
NM_001166060.1 Intron NP_001159532.1
NM_001166061.1 Intron NP_001159533.1
XM_017008034.1 Intron XP_016863523.1
XM_017008035.1 Intron XP_016863524.1

View Full Product Details