Product Details
- SNP ID
-
rs3777751
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:34804534 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGGTGATGCCAGGCAGAGGGAGC[C/T]GCATGAACAATGATACTGAGAAGCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
UHRF1BP1
PubMed Links
Gene Details
- Gene
- UHRF1BP1
- Gene Name
- UHRF1 binding protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017754.3 |
|
Intron |
|
|
NP_060224.3 |
View Full Product Details