Product Details

SNP ID
rs3783186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:42284254 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGATGCTCTAGGATCATTATTTGC[C/T]TTGATGCATTCCCAAAAGCAGAAAT
Phenotype
MIM: 604696
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKAP11 PubMed Links

Gene Details

Gene
AKAP11
Gene Name
A-kinase anchoring protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016248.3 Intron NP_057332.1
XM_005266247.1 Intron XP_005266304.1
XM_005266248.3 Intron XP_005266305.1
XM_005266249.3 Intron XP_005266306.1
XM_005266250.3 Intron XP_005266307.1
XM_011534903.2 Intron XP_011533205.1
XM_011534904.1 Intron XP_011533206.1
XM_011534905.1 Intron XP_011533207.1
XM_011534906.2 Intron XP_011533208.1
XM_017020381.1 Intron XP_016875870.1
XM_017020382.1 Intron XP_016875871.1
XM_017020383.1 Intron XP_016875872.1

View Full Product Details