Product Details

SNP ID
rs3784520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:101306544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCACCCTGCTGCTTGCTGGGCC[A/G]GAGCCCCCTTCCCTGCCCGTGTGTC
Phenotype
MIM: 167405
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100507472 PubMed Links

Gene Details

Gene
LOC100507472
Gene Name
uncharacterized LOC100507472
There are no transcripts associated with this gene.

Gene
PCSK6
Gene Name
proprotein convertase subtilisin/kexin type 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291309.1 Intron NP_001278238.1
NM_002570.4 Intron NP_002561.1
NM_138319.3 Intron NP_612192.1
NM_138322.3 Intron NP_612195.1
NM_138323.2 Intron NP_612196.1
NM_138324.2 Intron NP_612197.1
NM_138325.3 Intron NP_612198.2

View Full Product Details