Product Details

SNP ID
rs3788365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24004862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGTGATTCCCTTGTCTCTGTC[C/T]CTGCTCCTCAGAAACGCCCCTATCA
Phenotype
MIM: 604251
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CABIN1 PubMed Links

Gene Details

Gene
CABIN1
Gene Name
calcineurin binding protein 1
There are no transcripts associated with this gene.

Gene
GSTTP1
Gene Name
glutathione S-transferase theta pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details