Product Details
- SNP ID
-
rs3935504
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:178115308 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGGGGATGGGGTGGGCCAGCCTC[C/T]CCCAGCCCCTCTCTGGGCCCCCCTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
N4BP3
PubMed Links
Gene Details
- Gene
- N4BP3
- Gene Name
- NEDD4 binding protein 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015111.1 |
|
Intron |
|
|
NP_055926.1 |
View Full Product Details