Product Details

SNP ID
rs3794214
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120730442 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTCATTGAGAAGTGTCGCATTAG[C/T]CAGAGGGAGGCGGCGCTGTGTAGGG
Phenotype
MIM: 606885
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACADS PubMed Links

Gene Details

Gene
ACADS
Gene Name
acyl-CoA dehydrogenase, C-2 to C-3 short chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000017.3 Intron NP_000008.1
NM_001302554.1 Intron NP_001289483.1
Gene
MIR4700
Gene Name
microRNA 4700
There are no transcripts associated with this gene.

Gene
UNC119B
Gene Name
unc-119 lipid binding chaperone B
There are no transcripts associated with this gene.

View Full Product Details