Product Details

SNP ID
rs3796553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:155907133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACTCAATTGCCACAAAGTTCCCC[C/T]GGGAATTAGCTGTTACAGTAATATG
Phenotype
MIM: 191070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TDO2 PubMed Links

Gene Details

Gene
TDO2
Gene Name
tryptophan 2,3-dioxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005651.3 Intron NP_005642.1

View Full Product Details