Product Details
- SNP ID
-
rs3803794
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:21241687 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGGGTACTCAGGATGGTGACCCCT[G/C]CTGGTCCATGCTGTGACCCACAGCA
- Phenotype
-
- Polymorphism
- G/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NATD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77845899] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NATD1
- Gene Name
- N-acetyltransferase domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152914.2 |
1970 |
UTR 3 |
|
|
NP_690878.2 |
View Full Product Details