Product Details
- SNP ID
-
rs3806393
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
39
- Location
-
Chr.1:235329231 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- ACCAGGCCCTTCCCTTTGAAAAGTC[G/T]TTTGCATGGGATGTTCACTTCTTAG
- Phenotype
-
MIM: 609696
MIM: 606982
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ARID4B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147822910] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARID4B
- Gene Name
- AT-rich interaction domain 4B
- Gene
- GGPS1
- Gene Name
- geranylgeranyl diphosphate synthase 1
View Full Product Details