Product Details

SNP ID
rs3809009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:129375264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTGCGAGACTGCGAGTGGAGGG[C/T]TTGGGAAGCCGCGGTCTTCGGGAAA
Phenotype
MIM: 604823
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BARX2 PubMed Links

Gene Details

Gene
BARX2
Gene Name
BARX homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003658.4 28 Intron NP_003649.2
XM_011543043.1 28 Intron XP_011541345.1
XM_011543044.2 28 UTR 5 XP_011541346.1

View Full Product Details