Product Details

SNP ID
rs3814721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:71998226 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCAGAAACCCCAGGCAGCCTTGC[C/T]TCCTGACCTAACTCCTGGAGATGCT
Phenotype
MIM: 604113 MIM: 164009
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL18BP PubMed Links

Gene Details

Gene
IL18BP
Gene Name
interleukin 18 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039659.1 283 Intron NP_001034748.1
NM_001039660.1 283 Intron NP_001034749.1
NM_001145055.1 283 Intron NP_001138527.1
NM_001145057.1 283 Intron NP_001138529.1
NM_005699.3 283 Intron NP_005690.2
NM_173042.2 283 Intron NP_766630.2
NM_173044.2 283 Intron NP_766632.2
XM_017017059.1 283 Intron XP_016872548.1
XM_017017060.1 283 Intron XP_016872549.1
XM_017017061.1 283 Intron XP_016872550.1
XM_017017062.1 283 Intron XP_016872551.1
XM_017017063.1 283 UTR 5 XP_016872552.1
Gene
NUMA1
Gene Name
nuclear mitotic apparatus protein 1
There are no transcripts associated with this gene.

Gene
RNF121
Gene Name
ring finger protein 121
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300926.1 283 Intron NP_001287855.1
NM_018320.4 283 Intron NP_060790.2
XM_006718629.1 283 Intron XP_006718692.1
XM_006718630.1 283 Intron XP_006718693.1
XM_006718632.1 283 Intron XP_006718695.1
XM_011545150.1 283 Intron XP_011543452.1

View Full Product Details