Product Details

SNP ID
rs3820009
Assay Type
Functionally Tested
NCBI dbSNP Submissions
8
Location
Chr.1:2405893 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
GGGCCACTGGGCCATGCCCATCCCC[A/G]TCGGCATTTCTTTTCCTGTCCACAT
Phenotype
MIM: 602859 MIM: 612836
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PEX10 PubMed Links
Additional Information
For this assay, SNP(s) [rs75437503] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PEX10
Gene Name
peroxisomal biogenesis factor 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002617.3 Intron NP_002608.1
NM_153818.1 Intron NP_722540.1
XM_011541573.1 Intron XP_011539875.1
XM_011541576.1 Intron XP_011539878.1
Gene
PLCH2
Gene Name
phospholipase C eta 2
There are no transcripts associated with this gene.

Gene
RER1
Gene Name
retention in endoplasmic reticulum sorting receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007033.4 Intron NP_008964.3
XM_005244713.4 Intron XP_005244770.2
XM_006710306.2 Intron XP_006710369.1
XM_011540542.1 Intron XP_011538844.1
XM_011540543.1 Intron XP_011538845.1
XM_017000131.1 Intron XP_016855620.1
XM_017000132.1 Intron XP_016855621.1
XM_017000133.1 Intron XP_016855622.1
XM_017000134.1 Intron XP_016855623.1
XM_017000135.1 Intron XP_016855624.1

View Full Product Details