Product Details

SNP ID
rs3822617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:95466697 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTCTCTCCTTTGGTTTACCAGCT[A/G]GCCATTTTTACTTAGCCTTGCTTCT
Phenotype
MIM: 614589
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC37 PubMed Links

Gene Details

Gene
TTC37
Gene Name
tetratricopeptide repeat domain 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014639.3 Intron NP_055454.1

View Full Product Details