Product Details

SNP ID
rs3823446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:63647867 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTAAGGTTTTCCCTCACCATTCCC[G/T]AGAGTAAATAGTTCTCAGGTTTATG
Phenotype
MIM: 607789
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PHF3 PubMed Links

Gene Details

Gene
PHF3
Gene Name
PHD finger protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290259.1 Intron NP_001277188.1
NM_001290260.1 Intron NP_001277189.1
NM_015153.3 Intron NP_055968.1
XM_005248701.3 Intron XP_005248758.1
XM_005248702.3 Intron XP_005248759.1
XM_005248703.3 Intron XP_005248760.1
XM_006715426.3 Intron XP_006715489.1
XM_011535648.2 Intron XP_011533950.1
XM_017010626.1 Intron XP_016866115.1

View Full Product Details