Product Details

SNP ID
rs104894215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:88294111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCATTCTCACCCCAGCCGGTGCC[G/C]CAGCTGTTTTTAACAATCCAGTAAT
Phenotype
MIM: 602365
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
CTSC PubMed Links

Gene Details

Gene
CTSC
Gene Name
cathepsin C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114173.2 1402 Intron NP_001107645.1
NM_001814.5 1402 Missense Mutation TGC,TGG C429W NP_001805.3
NM_148170.4 1402 Intron NP_680475.1

View Full Product Details