Product Details
- SNP ID
-
rs121909598
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:208121728 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCCACTCTGGCATTCGTGGCCCCC[T/C]AGTCCTGGTAGCGCCTATAGTCCCC
- Phenotype
-
MIM: 123680
MIM: 123690
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CRYGC
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2305429] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CRYGC
- Gene Name
- crystallin gamma C
There are no transcripts associated with this gene.
- Gene
- CRYGD
- Gene Name
- crystallin gamma D
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006891.3 |
586 |
Nonsense Mutation |
TAG,TGG |
*157W |
NP_008822.2 |
- Gene
- LOC100507443
- Gene Name
- uncharacterized LOC100507443
There are no transcripts associated with this gene.
View Full Product Details