Product Details

SNP ID
rs120074145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:108134260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTGAAAGAAGCATACATGGGTA[A/G]TGTTCTACAAGGAGGTGAAGGACAA
Phenotype
MIM: 607809
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACAT1 PubMed Links

Gene Details

Gene
ACAT1
Gene Name
acetyl-CoA acetyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000019.3 337 Missense Mutation AAT,AGT N93S NP_000010.1
XM_006718835.3 337 Missense Mutation AAT,AGT N3S XP_006718898.1
XM_017017681.1 337 Missense Mutation AAT,AGT N3S XP_016873170.1
XM_017017682.1 337 Missense Mutation ATG,GTG M1V XP_016873171.1
XM_017017683.1 337 Missense Mutation ATG,GTG M1V XP_016873172.1

View Full Product Details