Product Details

SNP ID
rs4077176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:93064914 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTCCAGAGCTGGTCCTGGCTGCCT[C/T]GAGTGCATGGGAGGCCCTGGCGGTC
Phenotype
MIM: 616429
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SUSD3 PubMed Links

Gene Details

Gene
SUSD3
Gene Name
sushi domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287005.1 Intron NP_001273934.1
NM_001287006.1 Intron NP_001273935.1
NM_001287007.1 Intron NP_001273936.1
NM_001287008.1 Intron NP_001273937.1
NM_145006.3 Intron NP_659443.1
XM_011518358.1 Intron XP_011516660.1
XM_011518359.1 Intron XP_011516661.1
XM_011518361.2 Intron XP_011516663.1
XM_011518362.1 Intron XP_011516664.1
XM_017014448.1 Intron XP_016869937.1
XM_017014449.1 Intron XP_016869938.1
XM_017014450.1 Intron XP_016869939.1
XM_017014451.1 Intron XP_016869940.1

View Full Product Details