Product Details

SNP ID
rs4128984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:68662967 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGACTTAGCTCTGAGGAGGTAC[C/T]TAATGAATGAGTGAATAAATAAAGG
Phenotype
MIM: 300689
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STARD8 PubMed Links

Gene Details

Gene
STARD8
Gene Name
StAR related lipid transfer domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142503.2 Intron NP_001135975.1
NM_001142504.2 Intron NP_001135976.1
NM_014725.4 Intron NP_055540.2

View Full Product Details