Product Details
- SNP ID
-
hCV27536194
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:67545851 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGCACTTCGACCATTTCTGACAA[A/C]GCCAAGGAGTTGTGTAAGGCAGTGT
- Phenotype
-
MIM: 313700
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
AR
PubMed Links
Gene Details
- Gene
- AR
- Gene Name
- androgen receptor
There are no transcripts associated with this gene.
View Full Product Details