Product Details
- SNP ID
-
hCV27536271
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:37694503 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCTCAGCCCCCCAGCTGACCCTA[C/T]AGCTGTACATAAGTGTCATGCAGCA
- Phenotype
-
MIM: 314850
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
XK
PubMed Links
Gene Details
- Gene
- XK
- Gene Name
- X-linked Kx blood group
There are no transcripts associated with this gene.
View Full Product Details