Product Details

SNP ID
rs777638364
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.10:102830910 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
GGCCAGGATCTCACCTATACAGGAG[C/T]GAGGTCCTGCTCCGAAGGGCAAATA
Phenotype
MIM: 609300
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP17A1 PubMed Links

Gene Details

Gene
CYP17A1
Gene Name
cytochrome P450 family 17 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000102.3 1491 Missense Mutation CAC,CGC H440R NP_000093.1
Gene
CYP17A1-AS1
Gene Name
CYP17A1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
LOC107984264
Gene Name
transmembrane protein 108-like
There are no transcripts associated with this gene.

View Full Product Details