Product Details
- SNP ID
-
hCV27540391
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:44275899 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGCCTAGACTGGTGGCAGGAGGGG[C/T]CCATGTACCAGATCTACCCAAGGTC
- Phenotype
-
MIM: 104614
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SLC3A1
PubMed Links
Gene Details
- Gene
- SLC3A1
- Gene Name
- solute carrier family 3 member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000341.3 |
442 |
Missense Mutation |
CCC,TCC |
P122S |
NP_000332.2 |
View Full Product Details