Product Details

SNP ID
rs7186246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:80600162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGGCTTATTCACAAATTCAATA[C/T]ATTTTGAAATTATACTTCAAACGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDYL2 PubMed Links

Gene Details

Gene
CDYL2
Gene Name
chromodomain protein, Y-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152342.3 6407 UTR 3 NP_689555.2
XM_011522866.1 6407 UTR 3 XP_011521168.1
XM_011522867.2 6407 UTR 3 XP_011521169.1

View Full Product Details