Product Details

SNP ID
rs201144437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141223463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGAGCTTCAGCTTCCAAAATTAC[A/G]GCTGAGCTTCAGTTTTTCCACAAGA
Phenotype
MIM: 606339 MIM: 606340
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDHB13 PubMed Links

Gene Details

Gene
PCDHB13
Gene Name
protocadherin beta 13
There are no transcripts associated with this gene.

Gene
PCDHB14
Gene Name
protocadherin beta 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018934.3 1138 UTR 5 NP_061757.1

View Full Product Details