Product Details

SNP ID
rs1028880696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:131503775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGATGAGGAGGCCCCGAGGCTCG[G/T]CCAGGTTGGGGTCCCTGGGCTGTAA
Phenotype
MIM: 607423
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
POMT1 PubMed Links

Gene Details

Gene
POMT1
Gene Name
protein O-mannosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077365.1 Intron NP_001070833.1
NM_001077366.1 Intron NP_001070834.1
NM_001136113.1 Intron NP_001129585.1
NM_001136114.1 Intron NP_001129586.1
NM_007171.3 Intron NP_009102.3
Gene
PRRC2B
Gene Name
proline rich coiled-coil 2B
There are no transcripts associated with this gene.

View Full Product Details