Product Details

SNP ID
rs551395827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29973706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTCTAGCTCTTCTCCCCGCCTC[G/C]TCCGGCCTGGGGTCCGCGTGGCGGG
Phenotype
MIM: 613199
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC107987231 PubMed Links

Gene Details

Gene
LOC107987231
Gene Name
uncharacterized LOC107987231
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017023915.1 66 Missense Mutation CTC,GTC L29V XP_016879404.1
Gene
TAOK2
Gene Name
TAO kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252043.1 66 Intron NP_001238972.1
NM_004783.3 66 Intron NP_004774.1
NM_016151.3 66 Intron NP_057235.2
XM_011545982.1 66 UTR 5 XP_011544284.1
XM_011545983.2 66 UTR 5 XP_011544285.1
XM_011545984.1 66 UTR 5 XP_011544286.1
XM_011545985.1 66 UTR 5 XP_011544287.1
XM_011545986.2 66 UTR 5 XP_011544288.1
Gene
TMEM219
Gene Name
transmembrane protein 219
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083613.1 66 Intron NP_001077082.1
NM_194280.3 66 Intron NP_919256.1
XM_005255123.1 66 Intron XP_005255180.1

View Full Product Details