Product Details

SNP ID
rs73678525
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:12687622 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCTGAGTCCAAGGTTTCTCGTG[C/T]GGCAACTGCCAGAGCTGCTTGGAGA
Phenotype
MIM: 604786
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL4A PubMed Links

Gene Details

Gene
ARL4A
Gene Name
ADP ribosylation factor like GTPase 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037164.2 337 Intron NP_001032241.1
NM_001195396.1 337 UTR 5 NP_001182325.1
NM_005738.4 337 UTR 5 NP_005729.1
NM_212460.3 337 Intron NP_997625.1

View Full Product Details