Product Details

SNP ID
rs7980636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:39635924 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTGGAAATCTTACGAAGTTTGTA[C/G]CTATAAAATTAGTTGCCAAATTGCA
Phenotype
MIM: 601081
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ABCD2 PubMed Links

Gene Details

Gene
ABCD2
Gene Name
ATP binding cassette subfamily D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005164.3 Intron NP_005155.1
XM_011538027.2 Intron XP_011536329.1
XM_017018992.1 Intron XP_016874481.1
XM_017018993.1 Intron XP_016874482.1
XM_017018994.1 Intron XP_016874483.1
Gene
C12orf40
Gene Name
chromosome 12 open reading frame 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031748.3 Intron NP_001026918.2
NM_001319247.1 Intron NP_001306176.1
XM_005268806.3 Intron XP_005268863.1
XM_011538231.2 Intron XP_011536533.1

View Full Product Details