Product Details

SNP ID
rs45619231
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54115619 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACGTGAGTCCCTTTCCTCCTCG[A/C]GGCTTACCGCCTCTCTCCGCCTAGT
Phenotype
MIM: 603832 MIM: 606419 MIM: 609519
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NDUFA3 PubMed Links

Gene Details

Gene
NDUFA3
Gene Name
NADH:ubiquinone oxidoreductase subunit A3
There are no transcripts associated with this gene.

Gene
PRPF31
Gene Name
pre-mRNA processing factor 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015629.3 57 UTR 5 NP_056444.3
XM_006723137.3 57 UTR 5 XP_006723200.1
Gene
TFPT
Gene Name
TCF3 fusion partner
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321792.1 57 Intron NP_001308721.1
NM_013342.3 57 UTR 5 NP_037474.1
XM_005278261.1 57 Intron XP_005278318.1

View Full Product Details