Product Details

SNP ID
rs115721253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:101785373 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCACCCTAGGCCATTGCAATTGG[C/T]GATGTTCTCTCATCTCTCCTCTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NXPE3 PubMed Links

Gene Details

Gene
NXPE3
Gene Name
neurexophilin and PC-esterase domain family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134456.1 Intron NP_001127928.1
NM_145037.2 Intron NP_659474.1

View Full Product Details