Product Details

SNP ID
hCV27831307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55738474 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTCTTCACCGAGTCGCCGCCC[C/T]CCTACCCAGTCGACTCCCGTACCCC
Phenotype
MIM: 605281
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX4 PubMed Links

Gene Details

Gene
DDX4
Gene Name
DEAD-box helicase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142549.1 Intron NP_001136021.1
NM_001166533.1 Intron NP_001160005.1
NM_001166534.1 Intron NP_001160006.1
NM_024415.2 Intron NP_077726.1

View Full Product Details