Product Details

SNP ID
rs116077357
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:48222579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAGGAGTCCTGCAGGGGTTGGT[A/G]GTGGCAGTTGGGAGAAAAGGAGGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC5A9 PubMed Links

Gene Details

Gene
SLC5A9
Gene Name
solute carrier family 5 member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011547.2 1462 Intron NP_001011547.2
NM_001135181.1 1462 Intron NP_001128653.1
XM_011540924.2 1462 UTR 5 XP_011539226.1
XM_011540925.2 1462 UTR 5 XP_011539227.1
XM_011540926.2 1462 Intron XP_011539228.1
XM_011540927.1 1462 Intron XP_011539229.1
XM_011540928.1 1462 Intron XP_011539230.1
XM_011540929.2 1462 UTR 5 XP_011539231.1
XM_017000558.1 1462 UTR 5 XP_016856047.1

View Full Product Details