Product Details
- SNP ID
-
hCV27831748
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:132672524 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTGAGATGGAGATGAGGAGGAAC[A/C]CTCCGATTTTTTGGCAGGCCCTGTT
- Phenotype
-
MIM: 610450
MIM: 608789
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LYPD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114278512] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LYPD1
- Gene Name
- LY6/PLAUR domain containing 1
- Gene
- NCKAP5
- Gene Name
- NCK associated protein 5
View Full Product Details