Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014716.3 | 49 | Intron | NP_055531.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001002914.2 | 49 | Intron | NP_001002914.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001320435.1 | 49 | UTR 5 | NP_001307364.1 | ||
NM_001320436.1 | 49 | UTR 5 | NP_001307365.1 | ||
NM_198154.2 | 49 | UTR 5 | NP_937797.1 | ||
XM_017024565.1 | 49 | UTR 5 | XP_016880054.1 | ||
XM_017024566.1 | 49 | UTR 5 | XP_016880055.1 | ||
XM_017024567.1 | 49 | UTR 5 | XP_016880056.1 | ||
XM_017024568.1 | 49 | UTR 5 | XP_016880057.1 | ||
XM_017024569.1 | 49 | UTR 5 | XP_016880058.1 | ||
XM_017024570.1 | 49 | Intron | XP_016880059.1 | ||
XM_017024571.1 | 49 | UTR 5 | XP_016880060.1 |