Product Details

SNP ID
rs60606072
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.2:203239044 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
CTGGAGCGGCCGATCCGAGACGTGG[C/T]TCCCTGGGCGGCAGAACCATGTTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP20A1 PubMed Links

Gene Details

Gene
CYP20A1
Gene Name
cytochrome P450 family 20 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177538.2 71 UTR 5 NP_803882.1
XM_005246698.3 71 UTR 5 XP_005246755.1
XM_005246699.3 71 UTR 5 XP_005246756.1
XM_005246701.3 71 UTR 5 XP_005246758.1
XM_006712644.3 71 Intron XP_006712707.1
XM_006712645.3 71 UTR 5 XP_006712708.1
XM_017004555.1 71 UTR 5 XP_016860044.1

View Full Product Details