Product Details
- SNP ID
-
hCV27832274
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:179327885 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGTCTCCGCCTTTAGCTTGAAAA[C/G]TACCTAAAAATACTTCCAATGGTAG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF639
PubMed Links
Gene Details
- Gene
- ZNF639
- Gene Name
- zinc finger protein 639
View Full Product Details