Product Details

SNP ID
rs576019146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68272987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCACAGGAAACCTGCATGGCTTG[C/G]TGCCAGAGTGAGAGCAAAAGTGGTG
Phenotype
MIM: 605641
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC7A6 PubMed Links

Gene Details

Gene
SLC7A6
Gene Name
solute carrier family 7 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076785.2 Intron NP_001070253.1
NM_003983.5 Intron NP_003974.3
XM_011523433.1 Intron XP_011521735.1
XM_011523434.1 Intron XP_011521736.1
XM_011523438.1 Intron XP_011521740.1
XM_011523439.1 Intron XP_011521741.1
XM_017023851.1 Intron XP_016879340.1

View Full Product Details