Product Details

SNP ID
rs139365554
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:34889282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACGCGCTCGTGGGGAAAAGGCAG[C/G]GAGGGGGTGGTGTCCCCAGCCGGTT
Phenotype
MIM: 608994 MIM: 600772
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ANKS1A PubMed Links
Additional Information
For this assay, SNP(s) [rs74723861] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ANKS1A
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015245.2 10 UTR 5 NP_056060.2
XM_005248964.3 10 UTR 5 XP_005249021.1
XM_006715036.2 10 UTR 5 XP_006715099.1
XM_011514431.2 10 UTR 5 XP_011512733.1
XM_011514432.2 10 UTR 5 XP_011512734.1
XM_011514433.1 10 UTR 5 XP_011512735.1
XM_011514434.2 10 UTR 5 XP_011512736.1
XM_011514435.2 10 UTR 5 XP_011512737.1
XM_011514436.2 10 Intron XP_011512738.1
XM_011514437.2 10 Intron XP_011512739.1
XM_017010593.1 10 Intron XP_016866082.1
Gene
LOC105375030
Gene Name
uncharacterized LOC105375030
There are no transcripts associated with this gene.

Gene
TAF11
Gene Name
TATA-box binding protein associated factor 11
There are no transcripts associated with this gene.

View Full Product Details