Product Details

SNP ID
rs1048166138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:130001550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAGGGAGGGGTTAACTGAACTC[A/G]GGCTGGGGCTAGGGGGATGACATGA
Phenotype
MIM: 300688
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCORL1 PubMed Links

Gene Details

Gene
BCORL1
Gene Name
BCL6 corepressor-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184772.2 Intron NP_001171701.1
NM_021946.4 Intron NP_068765.3

View Full Product Details