Product Details
- SNP ID
-
rs141385203
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:28226856 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTAAAAGCTTATGCTCTTAGCAC[A/G]TTATGAATATATATATTTCCATTAT
- Phenotype
-
MIM: 602246
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TOB2P1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78793798] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TOB2P1
- Gene Name
- transducer of ERBB2, 2 pseudogene 1
There are no transcripts associated with this gene.
- Gene
- ZSCAN9
- Gene Name
- zinc finger and SCAN domain containing 9
View Full Product Details