Product Details

SNP ID
rs58795747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:41251142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAAGGCTGGTTAGAAGGTGCAA[A/G]TAGAAATCTACAGTGACAGTTATGT
Phenotype
MIM: 617010
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ULK4 PubMed Links

Gene Details

Gene
ULK4
Gene Name
unc-51 like kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322500.1 Intron NP_001309429.1
NM_001322501.1 Intron NP_001309430.1
NM_017886.3 Intron NP_060356.2
XM_011533874.2 Intron XP_011532176.1
XM_011533880.2 Intron XP_011532182.1
XM_017006711.1 Intron XP_016862200.1
XM_017006712.1 Intron XP_016862201.1

View Full Product Details