Product Details

SNP ID
rs928130840
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:209590474 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTGTATGGGAGGAGGAGTGGGC[A/G]GGAAAGATGTTTTCTCATAGCCTAG
Phenotype
MIM: 614994
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAMK1G PubMed Links

Gene Details

Gene
CAMK1G
Gene Name
calcium/calmodulin dependent protein kinase IG
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020439.2 Intron NP_065172.1

View Full Product Details