Product Details

SNP ID
rs56314564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5893749 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTGGTAGCTGCCTTATGGGTAG[A/G]CTTATTCACTTTTGTATCACTAATG
Phenotype
MIM: 607222
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD16 PubMed Links

Gene Details

Gene
ANKRD16
Gene Name
ankyrin repeat domain 16
There are no transcripts associated with this gene.

Gene
FBXO18
Gene Name
F-box protein, helicase, 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258452.1 491 Intron NP_001245381.1
NM_001258453.1 491 Intron NP_001245382.1
NM_032807.4 491 Intron NP_116196.3
NM_178150.2 491 Intron NP_835363.1
XM_011519748.2 491 Intron XP_011518050.1
XM_017016842.1 491 UTR 5 XP_016872331.1
XM_017016843.1 491 Intron XP_016872332.1
XM_017016844.1 491 Intron XP_016872333.1
XM_017016845.1 491 Intron XP_016872334.1
XM_017016846.1 491 Intron XP_016872335.1

View Full Product Details