Product Details

SNP ID
hCV27835373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102236456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGATGTGAGGGATGTGTGGCCCA[C/G]TCCCACTGGTATCAGGGTGGCAATC
Phenotype
MIM: 611815 MIM: 603698 MIM: 602669
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ELOVL3 PubMed Links

Gene Details

Gene
ELOVL3
Gene Name
ELOVL fatty acid elongase 3
There are no transcripts associated with this gene.

Gene
GBF1
Gene Name
golgi brefeldin A resistant guanine nucleotide exchange factor 1
There are no transcripts associated with this gene.

Gene
PITX3
Gene Name
paired like homeodomain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005029.3 Intron NP_005020.1

View Full Product Details