Product Details
- SNP ID
-
hCV27835440
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:108009295 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTCCGGCCTCCGGTCAAGGCCTG[A/G]CCGGGAGCGCCACGAATTCTCGCGT
- Phenotype
-
MIM: 601741
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CUL5
PubMed Links
Gene Details
- Gene
- CUL5
- Gene Name
- cullin 5
There are no transcripts associated with this gene.
View Full Product Details