Product Details

SNP ID
rs114221202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63698068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCTCCCCCATGGGCTGCAGGGG[A/C]ATTTATGATGCCCAACAGGTGGCAC
Phenotype
MIM: 602539 MIM: 608626
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LIMD2 PubMed Links

Gene Details

Gene
LIMD2
Gene Name
LIM domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030576.3 1157 UTR 3 NP_085053.1
XM_005257703.1 1157 UTR 3 XP_005257760.1
XM_005257705.3 1157 UTR 3 XP_005257762.1
XM_006722124.1 1157 UTR 3 XP_006722187.1
Gene
LOC729683
Gene Name
uncharacterized LOC729683
There are no transcripts associated with this gene.

Gene
MAP3K3
Gene Name
mitogen-activated protein kinase kinase kinase 3
There are no transcripts associated with this gene.

Gene
STRADA
Gene Name
STE20-related kinase adaptor alpha
There are no transcripts associated with this gene.

View Full Product Details