Product Details

SNP ID
rs58302291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:122884262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTAAGGTGCTTTCCTCCAAGGA[G/T]TTGATGATGCTCCTTGCAAACATTA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C11orf63 PubMed Links

Gene Details

Gene
C11orf63
Gene Name
chromosome 11 open reading frame 63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024806.3 Intron NP_079082.2
NM_199124.2 Intron NP_954575.1
XM_005271680.4 Intron XP_005271737.1
XM_006718915.2 Intron XP_006718978.1
XM_011543009.2 Intron XP_011541311.1
XM_017018338.1 Intron XP_016873827.1
XM_017018339.1 Intron XP_016873828.1
XM_017018340.1 Intron XP_016873829.1
XM_017018341.1 Intron XP_016873830.1
XM_017018342.1 Intron XP_016873831.1
XM_017018343.1 Intron XP_016873832.1
XM_017018344.1 Intron XP_016873833.1

View Full Product Details