Product Details

SNP ID
rs551348673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46492127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAGGACTGGGACAGTCTGGTCTG[G/T]ACTGTGAGGTCACACCCAAACCCTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PNMAL2 PubMed Links

Gene Details

Gene
PNMAL2
Gene Name
paraneoplastic Ma antigen family-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020709.2 3786 UTR 3 NP_065760.1
Gene
PPP5D1
Gene Name
PPP5 tetratricopeptide repeat domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205281.1 3786 Intron NP_001192210.1
XM_005258397.3 3786 Intron XP_005258454.1
XM_006722960.3 3786 Intron XP_006723023.1
XM_017026125.1 3786 Intron XP_016881614.1

View Full Product Details